Learn More
MSC Value Collection Sash-Cord Twine, Cotton, 7/32 in. Diameter, 100 ft.
Catalog No. 5000514186
Encompass
This sash-cord twine is constructed from natural cotton, designed for reliable utility and general-purpose applications.
  • Constructed from natural cotton
  • Diameter of 7/32 in.
  • Breaking strength of 235 lb.
  • Length per pound of 45 ft.
Catalog No. 50-005-14186 Supplier MSC Supplier No. 45890423
This sash-cord twine is constructed from natural cotton, designed for reliable utility and general-purpose applications.
  • Constructed from natural cotton
  • Diameter of 7/32 in.
  • Breaking strength of 235 lb.
  • Length per pound of 45 ft.