Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mopec™ Standard Elastane Cadaver Covers
SDP

Catalog No. 22445895 Shop All Mopec Products
Change view
Click to view available options
Color:
White
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
22-445-895 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-445-895 Supplier Mopec™ Supplier No. JZ027
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

These elastane cadaver covers are an ideal alternative to discretely cover cadavers being transported.

  • Elastane cadaver covers will require laundry service to be properly cleaned

Specifications

Product Type Cadaver Cover
Material Elastane
Height (English) 18 in.
Length (English) 79.4 in.
Width (English) 27.8 in.
Color White
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.