Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

XENOSEP TECHNOLOGIES STANDARD, EPA 1664A 4MG/ML

Catalog No. NC1906689
Encompass_Preferred

Standard, EPA 1664A 4mg/mL, 30 mL/pk

Catalog No. NC1906689 Supplier XENOSEP TECHNOLOGIES Supplier No. 26532
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Standard, EPA 1664A 4mg/mL, 30 mL/pk

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.