Learn More
Electron Microscopy Sciences Flexible Shaft Machine Standard Foot Speed Control 220 V
SDP
Catalog No. 50996807
Encompass_Preferred
Standard foot control is carbon resistance foot operated rheostat for infinity variable motor speeds. Electronic foot control is solid state with a plastic housing. For use with all flex shaft models except our full torque models.
Catalog No. 50-996-807 Supplier Electron Microscopy Sciences Supplier No. 6213811
Standard foot control is carbon resistance foot operated rheostat for infinity variable motor speeds. Electronic foot control is solid state with a plastic housing. For use with all flex shaft models except our full torque models.