Learn More
Fritsch Milling And Sizing Inc Standard Funnel for Long And Bulk Solids, P-19
SDP
Catalog No. 501534048
Encompass
This standard funnel for long- and bulk solids for PULVERISETTE 19 can be used to process most samples.
This standard funnel can be used to process free flowing materials, as well as to feed straw, film and other materials in their full length.
Alternatively a protected funnel with sample pusher is offered. Due to the sample pusher it enables a good handling and an outstanding material feeding.
Catalog No. 50-153-4048 Supplier Fritsch Milling And Sizing Inc Supplier No. 19.1550.00
This standard funnel for long- and bulk solids for PULVERISETTE 19 can be used to process most samples.
This standard funnel can be used to process free flowing materials, as well as to feed straw, film and other materials in their full length.
Alternatively a protected funnel with sample pusher is offered. Due to the sample pusher it enables a good handling and an outstanding material feeding.