Learn More
Perkin Elmer US LLC Standard Glass Baffled Cyclonic Spray Chamber for NexION 300/350
Catalog No. 502104810
Encompass_Preferred
This O-ring-free baffled cyclonic spray chamber, made of borosilicate glass, comes standard with NexION Q, X, and D models. It relies on momentum to efficiently and quickly remove the larger droplets from the sample mist decreasing any opportunity for occlusion or agglomeration of the smaller droplets. The result is higher efficiency, better precision, and faster washouts than other spray chamber designs. For use with ball joint injectors.
Catalog No. 50-210-4810 Supplier Perkin Elmer US LLC Supplier No. N8145014
This O-ring-free baffled cyclonic spray chamber, made of borosilicate glass, comes standard with NexION Q, X, and D models. It relies on momentum to efficiently and quickly remove the larger droplets from the sample mist decreasing any opportunity for occlusion or agglomeration of the smaller droplets. The result is higher efficiency, better precision, and faster washouts than other spray chamber designs. For use with ball joint injectors.