Learn More
Perkin Elmer US LLC Holmium Oxide Glass Filter
Supplier: Perkin Elmer US LLC N1010545
This filter ships with the Photometric and Wavelength Accuracy Reference Standards Set (B0507805), but with this part number, it can be ordered separately as a replacement or spare filter. It requires the Single Recalibration Kit (N1010540).