Learn More
Siemens Healthineers Standard Human Plasma (US) Calibration Standard
Catalog No. 23044735 Shop All Siemens Healthineers Products
Change view
Click to view available options
Quantity:
10 x 1 mL
Catalog No. Quantity
23-044-735 10 x 1 mL
1 options
This item is not returnable. View return policy
Catalog No. 23-044-735 Supplier Siemens Healthineers Supplier No. 10487098
Add to Cart
Add to Cart
This item is not returnable. View return policy
Standard Human Plasma for the calibration of coagulation and fibrinolysis tests.
Standard human plasma is citrated human normal pool plasma intended for the calibration of coagulation and fibrinolysis assays. Standard human plasma is calibrated against the respective WHO Standard, where available.
TRUSTED_SUSTAINABILITY
Specifications
Type Coagulation Control
For Use With (Application) Coagulation Testing
Quantity 10 x 1 mL