Learn More
Molecular Devices Molecular Devices SpectraMax iD5 Microplate Reader, Bottom Luminescence, 6-384 Well
Catalog No. 5000014666
Encompass_Preferred
The SpectraMax iD5 is a multi-mode microplate reader configurable with a bottom luminescence read mode for sensitive luminescence assays. It supports absorbance, fluorescence, luminescence, and time-resolved measurements across 6-384 well plates, and accepts optional injectors and micro-volume or blot accessories to expand workflow capability.
  • Five detection modes including absorbance, fluorescence, and luminescence.
  • Compatible with 6-384 well plate formats.
  • Optional injector system for on-plate reagent delivery.
  • Integrated touchscreen with onboard data acquisition software.
  • NFC-enabled filter identification for automated filter selection.
  • Temperature control up to 66°C and multiple shaking modes.
Catalog No. 50-000-14666 Supplier Molecular Devices Supplier No. ID5BL
Add to Cart
Add to Cart
Request a Quote
The SpectraMax iD5 is a multi-mode microplate reader configurable with a bottom luminescence read mode for sensitive luminescence assays. It supports absorbance, fluorescence, luminescence, and time-resolved measurements across 6-384 well plates, and accepts optional injectors and micro-volume or blot accessories to expand workflow capability.
  • Five detection modes including absorbance, fluorescence, and luminescence.
  • Compatible with 6-384 well plate formats.
  • Optional injector system for on-plate reagent delivery.
  • Integrated touchscreen with onboard data acquisition software.
  • NFC-enabled filter identification for automated filter selection.
  • Temperature control up to 66°C and multiple shaking modes.