Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hach Lange™ Standard Luminescent Probe

Catalog No. 501997742 Shop All Hach Company Products
Encompass
Change view
Click to view available options
Diameter (Metric):
15 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric)
50-199-7742 15 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-199-7742 Supplier Hach Lange™ Supplier No. LDO10101
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

LDO101 Standard Luminescent Dissolved Oxygen (LDO) probe with 1 m cable, Diameter: 15mm

LDO101 Standard Luminescent Dissolved Oxygen (LDO) probe with 1 m cable, Diameter: 15mm

TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) 15 mm
Type Standard Luminescent Probe
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.