Learn More
World Precision Instrument Standard Manipulator Arm, 2-Axes, Left handed
SDP
Catalog No. 502539192
Encompass
Easily Read Scales
The numerals on the scales are larger, and therefore more easily read. The scale lines are laser engraved, to allow finest possible permanent marking of scales on all 3 axes. Precise alignment with facing vernier scales gives accurate resolution to 0.1mm.
Catalog No. 50-253-9192 Supplier World Precision Instrument Supplier No. 505220
Easily Read Scales
The numerals on the scales are larger, and therefore more easily read. The scale lines are laser engraved, to allow finest possible permanent marking of scales on all 3 axes. Precise alignment with facing vernier scales gives accurate resolution to 0.1mm.