Learn More
Stoelting™ Standard Manipulator Arm
SDP
Catalog No. 10000135 Shop All Stoelting Co Products
Change view
Click to view available options
Model:
Add-on, Right
Left
Right
Standard, Left
Size:
2 axes
3 axes
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model Size
10-000-135 Right 2 axes
10-000-138 Left 2 axes
10-000-137 Standard, Left 3 axes
10-000-139 Add-on, Right 3 axes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 10-000-135 Supplier Stoelting™ Supplier No. 51602
Only null left
Add to Cart
Add to Cart
Standard Manipulator Arm controls medio-lateral and vertical positioning via lead screws, and antero-posterior movement via dovetail slide movement, with 80 mm of travel possible in each direction.
  • Replacement or add-on manipulator arms for existing stereotaxic
  • Universal joint allows the investigator to change the angle of the probe up to 90° in either the antero-posterior or medio-lateral planes
  • Locking mechanism will hold any angle position without slippage
  • Provides an absolute lock at 90° vertical
  • Easy-to-Read Scales
    • Numerals on the scales are larger
    • Scale lines are laser engraved to allow finest possible permanent marking of scales on all 3 axes
    • Precise alignment with facing vernier scales gives accurate resolution to 0.1 mm
  • Smooth Movements
    • Exclusive, triple lead screws on these manipulators allow the fastest positioning possible
    • Consistent with lining up the scales easily at a given coordinate
  • Swing joint allows the investigator to conveniently swing the manipulator arm and probe out of the way for performing a procedure – then reliably return the probe to the same point
  • Convenince for Electrophysiology - Integral brass bushings in the manipulator arm allow grounding directly to the closest metal on the manipulator arm—even the probe holder.
Specifications
For Use With (Application) Neuroscience
Size 2 axes
Model Right