Learn More
Alkali Scientific Standard Methods Agar 200mL, 250mL Glass 33 430, 1/Pk
SDP
Catalog No. 5000662760
Encompass_Preferred
Standard Methods Agar, 200mL in a 250mL Glass Bottle, 1/Pk is a general purpose microbiological medium formulated for the enumeration of heterotrophic bacteria in water, dairy, and food samples using standard plate count techniques. The non selective composition supports the growth of a wide variety of aerobic microorganisms, making it suitable for routine microbial monitoring and quality control analysis. Supplied in an autoclavable 250mL screw cap glass bottle containing 200mL of agar, the format is designed for convenient melting and use in pour plate procedures. Frequently used in laboratories following APHA and AWWA testing guidelines, this ready to melt medium provides reliable microbial recovery and consistent performance in environmental monitoring, regulatory testing, and food safety quality assurance programs.
Catalog No. 50-006-62760 Supplier Alkali Scientific Supplier No. SMAP01
Standard Methods Agar, 200mL in a 250mL Glass Bottle, 1/Pk is a general purpose microbiological medium formulated for the enumeration of heterotrophic bacteria in water, dairy, and food samples using standard plate count techniques. The non selective composition supports the growth of a wide variety of aerobic microorganisms, making it suitable for routine microbial monitoring and quality control analysis. Supplied in an autoclavable 250mL screw cap glass bottle containing 200mL of agar, the format is designed for convenient melting and use in pour plate procedures. Frequently used in laboratories following APHA and AWWA testing guidelines, this ready to melt medium provides reliable microbial recovery and consistent performance in environmental monitoring, regulatory testing, and food safety quality assurance programs.