Learn More
Alkali Scientific Standard Methods Agar 20mL, 10/Pk
SDP
Catalog No. 5000663122
Encompass_Preferred
Standard Methods Agar, 20mL Plates, 10/Pk is a non selective general purpose growth medium formulated for the enumeration of heterotrophic bacteria in water, dairy, and food samples using standard plate count methods. The balanced nutrient composition supports the growth of a broad range of aerobic and facultatively anaerobic microorganisms, enabling reliable microbial enumeration. Each plate contains 20mL of sterile agar that provides optimal depth and surface area for colony development and accurate counting. The ready to use plates simplify laboratory workflows by eliminating preparation steps and ensuring consistent results. Widely used in environmental monitoring, food safety laboratories, and public health testing programs, this medium supports routine microbial analysis and regulatory compliance testing procedures.
Catalog No. 50-006-63122 Supplier Alkali Scientific Supplier No. SMAT02
Standard Methods Agar, 20mL Plates, 10/Pk is a non selective general purpose growth medium formulated for the enumeration of heterotrophic bacteria in water, dairy, and food samples using standard plate count methods. The balanced nutrient composition supports the growth of a broad range of aerobic and facultatively anaerobic microorganisms, enabling reliable microbial enumeration. Each plate contains 20mL of sterile agar that provides optimal depth and surface area for colony development and accurate counting. The ready to use plates simplify laboratory workflows by eliminating preparation steps and ensuring consistent results. Widely used in environmental monitoring, food safety laboratories, and public health testing programs, this medium supports routine microbial analysis and regulatory compliance testing procedures.