Learn More
Bioworld Standard Methods Agar, 500 g
Catalog No. 50254372
Encompass_Preferred
Standard Methods AgarUsed for obtaining microbial plate counts from milk and dairy products, foods, water and other materials of sanitary importanceMade according to the American Public Health Association (APHA) formulationpH: 7.0Working Concentration: 23.5 g/LDehydrated Appearance: light beige, homogeneous powderPrepared Appearance: light amber agarSoluble in boiling waterContains (g/L): Pancreatic Digest of Casein-5.0g Yeast Extract-2.5g Glucose-1.0g Agar-15.0g
Catalog No. 50-254-372 Supplier Bioworld Supplier No. 306200781
Standard Methods AgarUsed for obtaining microbial plate counts from milk and dairy products, foods, water and other materials of sanitary importanceMade according to the American Public Health Association (APHA) formulationpH: 7.0Working Concentration: 23.5 g/LDehydrated Appearance: light beige, homogeneous powderPrepared Appearance: light amber agarSoluble in boiling waterContains (g/L): Pancreatic Digest of Casein-5.0g Yeast Extract-2.5g Glucose-1.0g Agar-15.0g