Learn More
Alkali Scientific Standard Methods Agar (PCA) 10mL 60mm, 10/Pk
SDP
Supplier:  Alkali Scientific SMAP03
Encompass_Preferred
Standard Methods Agar (PCA), 10mL, 60mm Plate, 10/Pk is a non selective microbiological medium formulated for the enumeration of viable aerobic bacteria in food, dairy, and water samples using plate count methods. Also known as Plate Count Agar (PCA), it is widely recommended by regulatory organizations including APHA and FDA for routine microbial monitoring and quality control testing. Each 60mm Petri dish contains 10mL of sterile agar, providing a compact format suitable for low volume sampling, field testing, and laboratories managing high sample throughput. The ready to use plates ensure consistent medium depth and reliable colony development. This medium supports accurate microbial recovery and enumeration in environmental monitoring, food safety testing, and regulated laboratory workflows.
Catalog No. 50-006-62834