Learn More
V&P Scientific Inc MICROPLATE, PTFE, 96 Standard Round Well, Round Bottom, 0.35mL
Catalog No. NC2176881
Encompass
MICROPLATE, 96 Well Format, 8mm Diameter, 0.35mL Volume, Round Well, Round Bottom, SLAS Footprint, PTFE
This item is not returnable. View return policy
Catalog No. NC2176881 Supplier V&P Scientific Inc Supplier No. VP 416PTFE
MICROPLATE, 96 Well Format, 8mm Diameter, 0.35mL Volume, Round Well, Round Bottom, SLAS Footprint, PTFE