Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Standard Mixture of Fatty Acid Methyl Esters, TCI America™
SDP

Catalog No. S04605ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S04605ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S04605ML Supplier TCI America Supplier No. S04605ML
Add to Cart
Add to Cart

(consists of Methyl Decanoate, Laurate, Myristate, Palmitate and Stearate), [Standard Material for GC]

Chemical Identifiers

MDL Number MFCD00197827
Synonym Fatty Acid Methyl Ester Mixture

Specifications

Physical Form Liquid
Color Colorless
Quantity 5 mL
Chemical Name or Material Standard Mixture of Fatty Acid Methyl Esters (consists of Methyl Decanoate, Laurate, Myristate, Palm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.