Learn More
Waters Corp Waters Corporation Needle Assembly, 15μL, Standard, For 2707 Autosampler
Catalog No. NC1680058 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This 15μL Needle Assembly is a standard replacement component for the 2707 Autosampler. It is designed for use with HPLC System Type and specifically with the Investigator SFC System. Annual replacement is recommended.
  • Standard needle for the 2707 Autosampler.
  • Designed for HPLC system type.
  • Compatible with Investigator SFC System.
  • Recommended annual replacement.
  • Component for 2707 Sample Manager.
  • One unit per package.
Catalog No. NC1680058 Supplier Waters Corp Supplier No. 700003842
This 15μL Needle Assembly is a standard replacement component for the 2707 Autosampler. It is designed for use with HPLC System Type and specifically with the Investigator SFC System. Annual replacement is recommended.
  • Standard needle for the 2707 Autosampler.
  • Designed for HPLC system type.
  • Compatible with Investigator SFC System.
  • Recommended annual replacement.
  • Component for 2707 Sample Manager.
  • One unit per package.