Learn More
SPEX CERTIPREP LLC 3P STANDARD NITRITE 100PPM 250ML
Catalog No. NC9777570
Encompass_Preferred
(HPS by Zeptometrix) 100 ug/mL Nitrite from NaNO2 in water, 99.99+% (metals basis); Single Component IC Standard, 12 Month Expiry Date. Prepared from high-purity salts in 18-megaohm water, packaged in HDPE bottles.
Catalog No. NC9777570 Supplier SPEX CERTIPREP LLC 3P Supplier No. ICN250
(HPS by Zeptometrix) 100 ug/mL Nitrite from NaNO2 in water, 99.99+% (metals basis); Single Component IC Standard, 12 Month Expiry Date. Prepared from high-purity salts in 18-megaohm water, packaged in HDPE bottles.