Learn More
Cole-Parmer Oakton Traceable Conductivity and TDS Standard, Individually-Tested, 1000 µS; 500 mL
Catalog No. 501961267 Shop All Cole-Parmer Products
Encompass_Preferred
All Oakton Traceable conductivity standards are NIST-traceable reference materials Supplied with a calibration report from an ISO 17034 and ISO 17025 accredited laboratory These standard solutions let you calibrate conductivity instruments to ensure accuracy If unopened, shelf life is one year; once opened, shelf life will vary by solution value. Standards have detailed labels: KCl, NaCl, and 442 conversions, temperature corrections, and certified values referenced to 25C. Average accuracy at 25C is 0.25 microsiemens for 1, 5, 10 microsiemen (S) solutions, or 0.25% for all other solutions. The tested uncertainty is shown on the certificate, whichever is greater.
Catalog No. 50-196-1267 Supplier Cole-Parmer Supplier No. 0065248
All Oakton Traceable conductivity standards are NIST-traceable reference materials Supplied with a calibration report from an ISO 17034 and ISO 17025 accredited laboratory These standard solutions let you calibrate conductivity instruments to ensure accuracy If unopened, shelf life is one year; once opened, shelf life will vary by solution value. Standards have detailed labels: KCl, NaCl, and 442 conversions, temperature corrections, and certified values referenced to 25C. Average accuracy at 25C is 0.25 microsiemens for 1, 5, 10 microsiemen (S) solutions, or 0.25% for all other solutions. The tested uncertainty is shown on the certificate, whichever is greater.