Learn More
Cole-Parmer Oakton Traceable Conductivity and TDS Standard, Batch-Tested, 1413 µS; 500 mL
Catalog No. 501961262 Shop All Cole-Parmer Products
Encompass_Preferred
Traceable conductivity standards are Certified Reference Materials, making one of the most accurate available
Catalog No. 50-196-1262 Supplier Cole-Parmer Supplier No. 0065230
Traceable conductivity standards are Certified Reference Materials, making one of the most accurate available