Learn More
Cole-Parmer Oakton Traceable One-Shot™ Conductivity and TDS Standard, Assortment; 6 x 100 mL Vials
Catalog No. 501961722 Shop All Cole-Parmer Products
Encompass_Preferred
Oakton Traceable One-Shot™ conductivity and TDS standards include detailed KCl, NaCl, and 442 conversions, temperature corrections, and certified values referenced to 25C. Average accuracy at 25C is 0.25 microsiemens for 1, 5, 10 microsiemen (S) solutions, or 0.25% for all other solutions. The tested uncertainty is shown on the certificate, whichever is greater. Conductivity options range from 1 to 200,000 S/cm. Batch-tested solutions are lot certified; a few bottles from a single lot are analyzed
Catalog No. 50-196-1722 Supplier Cole-Parmer Supplier No. 0065274
Oakton Traceable One-Shot™ conductivity and TDS standards include detailed KCl, NaCl, and 442 conversions, temperature corrections, and certified values referenced to 25C. Average accuracy at 25C is 0.25 microsiemens for 1, 5, 10 microsiemen (S) solutions, or 0.25% for all other solutions. The tested uncertainty is shown on the certificate, whichever is greater. Conductivity options range from 1 to 200,000 S/cm. Batch-tested solutions are lot certified; a few bottles from a single lot are analyzed