Learn More
Fine Science Tools Standard Pattern Forceps, Stainless Steel, 18 cm
SDP
Catalog No. NC1147044
Encompass
Tip Shape: Straight/ Tips: Serrated/ Tip Dimensions: 3.7 x 1.85mm/ Alloy / Material: Stainless Steel/ Length: 18cm
Catalog No. NC1147044 Supplier Fine Science Tools Supplier No. 1100018
Tip Shape: Straight/ Tips: Serrated/ Tip Dimensions: 3.7 x 1.85mm/ Alloy / Material: Stainless Steel/ Length: 18cm