Learn More
Fine Science Tools Surgical Scissors, Large Loops, Stainless Steel, 14.5 cm
SDP
Catalog No. NC9197296
Encompass
Tip Shape: Straight/ Tips: Sharp-Blunt/ Cutting Edge: 45mm/ Alloy / Material: Stainless Steel/ Length: 14.5cm/ Serrated: No/ Feature: Large Loops
Catalog No. NC9197296 Supplier Fine Science Tools Supplier No. 1410114
Tip Shape: Straight/ Tips: Sharp-Blunt/ Cutting Edge: 45mm/ Alloy / Material: Stainless Steel/ Length: 14.5cm/ Serrated: No/ Feature: Large Loops