Learn More
Andwin Scientific STANDARD PETRI DISHES 100 X 1
SDP
Catalog No. 50820904
Encompass_Preferred
The 100 x 15 mm PS petri dish is constructed with biomedical grade crystal polystyrene that is exceptionally clear. With flat bottoms, this product prevents condensation build-up caused by expensive media and venting ribs. This product is sterilized with the use of gamma radiation. The dishes are also stackable.
Catalog No. 50-820-904 Supplier Andwin Scientific Supplier No. 86090010
The 100 x 15 mm PS petri dish is constructed with biomedical grade crystal polystyrene that is exceptionally clear. With flat bottoms, this product prevents condensation build-up caused by expensive media and venting ribs. This product is sterilized with the use of gamma radiation. The dishes are also stackable.