Learn More
Andwin Scientific STANDARD PETRI DISHES 60 X 15
SDP
Catalog No. 50820905
Encompass
The 60 x 15 mm PS petri dish is constructed with biomedical grade crystal polystyrene that is exceptionally clear. With flat bottoms, this product prevents condensation build-up caused by expensive media and venting ribs. This product is sterilized with the use of gamma radiation. The dishes are also stackable.
Catalog No. 50-820-905 Supplier Andwin Scientific Supplier No. 86090160
The 60 x 15 mm PS petri dish is constructed with biomedical grade crystal polystyrene that is exceptionally clear. With flat bottoms, this product prevents condensation build-up caused by expensive media and venting ribs. This product is sterilized with the use of gamma radiation. The dishes are also stackable.