Learn More
SPEX CERTIPREP LLC 3P Drinking Water Standard Plate Count
Catalog No. NC0441179
Encompass_Preferred
(NSI by Zeptometrix) A single sample for quantitative determination of Heterotrophic Bacteria by plate count or MPN methods. Sample derived from E. faecalis( NCTC 775) at an activity level of 5-500mpn/mL after dilution in supplied hydration fluid. Sample taken from previous Interlab PT study and accompanying COA contains study statistics. Stores at -10C to -20C.
Catalog No. NC0441179 Supplier SPEX CERTIPREP LLC 3P Supplier No. MICQC3
(NSI by Zeptometrix) A single sample for quantitative determination of Heterotrophic Bacteria by plate count or MPN methods. Sample derived from E. faecalis( NCTC 775) at an activity level of 5-500mpn/mL after dilution in supplied hydration fluid. Sample taken from previous Interlab PT study and accompanying COA contains study statistics. Stores at -10C to -20C.