Learn More
Mopec™ Standard Platform Scale
SDP
Catalog No. 22445482 Shop All Mopec Products
Change view
Click to view available options
Capacity:
2200 g
Catalog No. Capacity
22-445-482 2200 g
1 options
Catalog No. 22-445-482 Supplier Mopec™ Supplier No. BB065
Add to Cart
Add to Cart
Ideal for laboratory and industrial applications, the Platform Scale features an easy to navigate color touchscreen display and slim design, allowing for stackable storage. Geared for high performance with fast stabilization time and high resolution weighing results.
  • Includes overload protection and expanded connectivity options
  • Multiple weighing units
  • USB Host, USB Device or RS232 connectivity
  • Battery or AC Power
Specifications
Capacity 2200 g
Display Type Digital