Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Grainger Standard Platform Truck 2000 l

Catalog No. 502771870
Encompass_Preferred

Standard Platform Truck 2000 lb 48 In L

Catalog No. 50-277-1870 Supplier Grainger Supplier No. 13X473
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Standard Platform Truck 2000 lb 48 In L

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.