Learn More
World Precision Instrument Standard Probe Holder/Corner Mount
SDP
Catalog No. 502521329
Encompass
Holds probes 0.3 mm to 1.5 mm OD.
Catalog No. 50-252-1329 Supplier World Precision Instrument Supplier No. 502210
Holds probes 0.3 mm to 1.5 mm OD.