Learn More
Stoelting™ Standard Probe Holder - Side Clamp
SDP
Catalog No. 10000382 Shop All Stoelting Co Products
Change view
Click to view available options
For Use With (Application):
Neuroscience
Catalog No. For Use With (Application)
10-000-382 Neuroscience
1 options
Catalog No. 10-000-382 Supplier Stoelting™ Supplier No. 516362
 Restricted Item
Add to Cart
Add to Cart
Stoelting Standard Probe Holder Side Clamp includes an 11mm square acrylic block connected to a 19.5 cm standard shaft (6 mm diameter) with grooves spaced 1 mm apart
Specifications
For Use With (Application) Neuroscience