Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Accumax Lab Devices Standard Profile Optical 384 Well PCR Plate, Alpha Numeric Printed

Catalog No. 02945146
Change view
Click to view available options
Color:
Natural
White
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
02-945-146 White
02-945-140 Natural
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 02-945-146 Supplier Accumax Lab Devices Supplier No. 335816
Add to Cart
Add to Cart

PCR plates designed to make lab work easier while providing exceptional heat transfer and precise, error-free results.

  • Developed from expertise in quality liquid handling equipment.
  • Various design aspects help make lab work easier.
  • Ultra-thin but robust walls provide exceptional heat transfer.
  • Uniform wall thickness supports precise and error-free results.
  • Manufactured to consistent quality standards.
  • Free from RNase/DNase/Human DNA/PCR inhibitors
  • Ultra thin yet robust walls ensures maximum heat transfer
  • Specially designed smaller bulk packs help avoid contamination by multiple users
  • Non-reactive internal surface gives consistent and error free results
  • Manufactured in 100 K clean room with USFDA approved resins

Specifications

Barcode No Barcode
Format Standard
Array 24 x 16
Color White
Disposable Single-use
Product Type PCR Well Plate
Skirt Style Fully Skirted
Material Polypropylene
Volume (Metric) Well 0.05 mL
No. of Wells 384
Sterility Sterile
Well Shape Conical Bottom (V-well)
Dnase Free Yes
For Use With (Application) PCR
Quantity 50
RNase Free Yes
Fit for Purpose Sterile and PCR Ready
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.