Learn More
Perkin Elmer US LLC Standard PVC MP2 Two-Stop Peristaltic Pump Tubing - 1.14 mm I.D., Red/Red, Pkg. 12
Catalog No. 502104943
Encompass_Preferred
MP2 Two-Stop Peristaltic Pump Tubing. Product Features: PVC Tubing, 1.14mm I.D., Bridge Distance 72mm, Red-Red, 12 Pack.
Catalog No. 50-210-4943 Supplier Perkin Elmer US LLC Supplier No. N8145158
MP2 Two-Stop Peristaltic Pump Tubing. Product Features: PVC Tubing, 1.14mm I.D., Bridge Distance 72mm, Red-Red, 12 Pack.