Learn More
Perkin Elmer US LLC Standard PVC MP2 Two-Stop Peristaltic Pump Tubing - 0.76 mm I.D., Black/Black, Pkg. 12
Catalog No. 502104925
Encompass_Preferred
MP2 Two-Stop Peristaltic Pump Tubing. Product Features: PVC Tubing, 0.76mm I.D., Bridge Distance 72mm, Black-Black, 12 Pack.
Catalog No. 50-210-4925 Supplier Perkin Elmer US LLC Supplier No. N8145153
MP2 Two-Stop Peristaltic Pump Tubing. Product Features: PVC Tubing, 0.76mm I.D., Bridge Distance 72mm, Black-Black, 12 Pack.