Learn More
Perkin Elmer US LLC Standard PVC Two-Stop Peristaltic Pump Tubing - 2.06 mm I.D., Purple/Purple, Pkg. 12
Catalog No. 502105304
Encompass_Preferred
This non-flared PVC Two-Stop Pump Tubing is for use with the standard pump in Optima, Avio, and NexION 300/350 series instruments. This tubing ships as a package of 12.
Catalog No. 50-210-5304 Supplier Perkin Elmer US LLC Supplier No. N0777561
This non-flared PVC Two-Stop Pump Tubing is for use with the standard pump in Optima, Avio, and NexION 300/350 series instruments. This tubing ships as a package of 12.