Learn More
Cayman Chemical Standard qPCR Enzyme Mastermix, 1 ea
SDP
Catalog No. 502942965
Encompass_Preferred
The qPCR enzyme mastermix, minimizes primer dimer artifacts and optimized Cq performance. Custom formulations for qPCRassays.com from Empirical Biosciences (Fortis). Organism Type: Enzyme mastermix Instruments: Any instrument
Catalog No. 50-294-2965 Supplier Cayman Chemical Supplier No. 407051 EA
The qPCR enzyme mastermix, minimizes primer dimer artifacts and optimized Cq performance. Custom formulations for qPCRassays.com from Empirical Biosciences (Fortis). Organism Type: Enzyme mastermix Instruments: Any instrument