Learn More
BINDER™ Standard Rack
SDP
Catalog No. 502875755 Shop All BINDER Inc. Products
Change view
Click to view available options
For Use With (Equipment):
KB 65
KB PRO/KBF/KBF PRO 1600
KB/KB PRO 1060, KBF/KBF PRO 1060
KB/KB PRO 130, KBF/KBF PRO 130
KB/KB PRO 720, KBF/KBF PRO 720
KB/KB PRO/KBF/KBF PRO 260/470
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
50-287-5755 KB 65
50-287-5644 KB/KB PRO 130, KBF/KBF PRO 130
50-287-5709 KB/KB PRO/KBF/KBF PRO 260/470
50-287-5682 KB/KB PRO 720, KBF/KBF PRO 720
50-287-5731 KB/KB PRO 1060, KBF/KBF PRO 1060
50-287-5615 KB PRO/KBF/KBF PRO 1600
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 50-287-5755 Supplier BINDER™ Supplier No. 80122388
Add to Cart
Add to Cart
Shelf rack in-set with pair of retaining U-rails, for use with climate chambers and cooling incubator chambers.
  • Shelf rack made of stainless steel
  • Includes set of U-rails
Specifications
For Use With (Equipment) KB 65
Material Stainless Steel
Type Rack