Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Spectrum™ Standard RC Dialysis Tubing
SDP

Catalog No. 08670248 Shop All Spectrum Medical Industries Products
Change view
Click to view available options
Diameter (Metric):
6.4 mm
16 mm
20.4 mm
48 mm
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric)
08-670-248 48 mm
08-670-245 6.4 mm
08-670-4B 16 mm
08-670-246 20.4 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 08-670-248 Supplier Spectrum™ Supplier No. 132709
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Multipurpose dialysis tubing is ideal for general dialysis for a variety of applications and volumes

  • Membrane grade
  • Membrane type: RC
  • Molecular weight cut-off: 12 to 14K

Specifications

Product Type Dialysis Membrane
MWCO 12 to 14 kDa
Material Regenerated Cellulose
Diameter (Metric) 48 mm
Diameter (English) 1.88 in.
Length (English) 50 ft.
Length (Metric) 15 m
Capacity (Metric) 18 mL
Quantity 50 Pack
For Use With (Application) Some tubing rolls do not contain glycerin and do not require soaking prior to use
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.