Learn More
Hach Company Standard redox solution
Supplier: Hach Company LZW9402.99
Standard redox solution-220 mV-125mlStandard for calibrating and checking accuracy of ORP/Redox electrodes 125mL bottle