Learn More
Starna Cells Inc Rectangular Spectrophotometer Cells, 100 mm
SDP
Catalog No. NC1789824
Encompass_Preferred
Standard window and wall thickness (1.25mm). Window material Optical Glass, useable range: 334 to 2500nm. Can be used with spectrophotometers and colorimeters.
Catalog No. NC1789824 Supplier Starna Cells Inc Supplier No. 21G100
Standard window and wall thickness (1.25mm). Window material Optical Glass, useable range: 334 to 2500nm. Can be used with spectrophotometers and colorimeters.