Learn More
Fisherbrand™ Standard Rotor Cover
GREENER_CHOICE
Catalog No. 12006910 Shop All Fisherbrand Products
Change view
Click to view available options
For Use With (Equipment):
Mini-Centrifuge
Catalog No. For Use With (Equipment)
12-006-910 Mini-Centrifuge
1 options
Catalog No. 12-006-910
Add to Cart
Add to Cart
Provides protection from spinning contents in centrifuge. Fisherbrand™ Standard Rotor Cover should be placed on Mini Centrifuge prior to use.
  • For use with Fisherbrand™ Mini-Centrifuge
  • Holds: 12 mL x 1.5 mL/2.0 mL Tube
  • Limits noise and aspiration that may escape from tubes
Specifications
Material Plastic
Type Rotor Cover
Capacity None
Max. RCF None
Max. Speed None
For Use With (Equipment) Mini-Centrifuge
Biocontainment No
Autoclavable Not Autoclavable