Learn More
Fritsch Milling And Sizing Inc Standard Rotor P-19 Cpl. with V-Cutting Edges, Stainl. Steel
SDP
Catalog No. 501534476
Encompass
Standard rotor with V-cutting edges and fixed knives made of tool steel
for gentle size reduction through pure cutting action.
Catalog No. 50-153-4476 Supplier Fritsch Milling And Sizing Inc Supplier No. 45.5100.00
Add to Cart
Add to Cart
Request a Quote
Standard rotor with V-cutting edges and fixed knives made of tool steel
for gentle size reduction through pure cutting action.