Learn More
Perkin Elmer US LLC Standard Sample Pan Crimper Head - Replacement
Catalog No. 502117257
Encompass_Preferred
The Standard Sample Pan Crimper Press is used to crimp covers on standard DSC pans of aluminum, gold, and copper.
Catalog No. 50-211-7257 Supplier Perkin Elmer US LLC Supplier No. 02191171
The Standard Sample Pan Crimper Press is used to crimp covers on standard DSC pans of aluminum, gold, and copper.