Learn More
Perkin Elmer US LLC Standard Sample Pan Crimper Press
Catalog No. 502112127
Encompass_Preferred
Used to crimp covers on standard DSC pans of aluminum, gold and copper. Design incorporates a replaceable crimper head.
Catalog No. 50-211-2127 Supplier Perkin Elmer US LLC Supplier No. 02190048
Used to crimp covers on standard DSC pans of aluminum, gold and copper. Design incorporates a replaceable crimper head.