Learn More
Glass Expansion Standard Sample Tubing 1/16" OD x 0.30mm ID x 10m long
SDP
Catalog No. 5000555706
Encompass_Preferred
Standard Sample Tubing 1/16" OD x 0.30mm ID x 10m long Sourced from Glass Expansion
Catalog No. 50-005-55706 Supplier Glass Expansion Supplier No. STT1630
Standard Sample Tubing 1/16" OD x 0.30mm ID x 10m long Sourced from Glass Expansion