Learn More
Glass Expansion Standard Sample Tubing 1/16" OD x 1.0mm ID x 10m long
SDP
Catalog No. 5000555671
Encompass_Preferred
Standard Sample Tubing 1/16" OD x 1.0mm ID x 10m long Sourced from Glass Expansion
Catalog No. 50-005-55671 Supplier Glass Expansion Supplier No. STT16100
Standard Sample Tubing 1/16" OD x 1.0mm ID x 10m long Sourced from Glass Expansion