Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Standard Samples for HPLC, Set 2: Methyl Paraben in Methanol/Water, LiChroTest™, MilliporeSigma™

Catalog No. M1191580001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 Units
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1191580001 5 Units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1191580001 Supplier MilliporeSigma Supplier No. 1191580001
Add to Cart
Add to Cart

Chemical Identifiers

Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
IUPAC Name methanol
SMILES CO

Specifications

Packaging Glass ampoule
Odor Methanolic
Physical Form Liquid
UN Number UN 1230
Color Colorless
Quantity 5 Units
Chemical Name or Material Standard Samples for HPLC, Set 2
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.