Learn More
Ampco Standard Screwdriver
SDP
Catalog No. 19174271 Shop All Ampco Metal Inc. Products
Change view
Click to view available options
Length (English):
6 -1/2 in.
8 -1/2 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English)
19-174-271 6 -1/2 in.
19-174-272 8 -1/2 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-174-271 Supplier Ampco Supplier No. S35
Add to Cart
Add to Cart
Flathead screwdriver with fluted grip handle. Includes a lifetime warranty.
  • Cellulose acetate fluted grip handle.
  • Beryllium copper material.
  • Made in the USA.,
  • 4 mm (5/32 in.) tip size (slotted blade).
Specifications
Product Type Standard Screwdriver
Length (English) 6 -1/2 in.
Tip Style Standard