Learn More
R&D Systems Standard scWest Kit
SDP
Catalog No. NC1429455
Encompass
Kit of 8 scWest chips (standard well size) and all wash, lysis/run, suspension, and blocking buffers to run 8 chips.
Catalog No. NC1429455 Supplier R&D Systems Supplier No. K600/DEL
Customers require a valid business account associated to their profile to access pricing and ordering options.
Kit of 8 scWest chips (standard well size) and all wash, lysis/run, suspension, and blocking buffers to run 8 chips.